missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81, Rabbit anti-Human,Chimpanzee,Goat,Sheep, Clone: 1D6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81096-0.025MG
This item is not returnable.
View return policy
Description
CD81 Monoclonal specifically detects CD81 in Human, Goat, Chimpanzee, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Functional.
Specifications
| CD81 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Functional | |
| CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody is specific for human CD81, which is expressed on most tissues with the exception of red blood cells and platelets. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1D6 | |
| Unconjugated | |
| PBS | |
| CD81 | |
| This antibody was produced by immunizing mice with 5A6-aggregated OCI-LY8 B cells. | |
| 0.025 mg | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 975 | |
| Human, Chimpanzee, Goat, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction