missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD81, Rabbit anti-Human,Chimpanzee,Goat,Sheep, Clone: 1D6, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 281.00 - € 583.00
Specifications
| Antigen | CD81 |
|---|---|
| Clone | 1D6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Functional |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18032554
|
Novus Biologicals
NBP2-81096-0.025MG |
0.025 mg |
€ 281.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18256269
|
Novus Biologicals
NBP2-81096-0.2MG |
0.2 mg |
€ 583.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD81 Monoclonal specifically detects CD81 in Human, Goat, Chimpanzee, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Functional.Specifications
| CD81 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| PBS | |
| 975 | |
| This antibody was produced by immunizing mice with 5A6-aggregated OCI-LY8 B cells. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1D6 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Functional | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Chimpanzee, Goat, Sheep | |
| CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7 | |
| CD81 | |
| IgG κ | |
| Protein A purified | |
| This antibody is specific for human CD81, which is expressed on most tissues with the exception of red blood cells and platelets. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title