Learn More
CD81, Rabbit anti-Human,Chimpanzee,Goat,Sheep, Clone: 1D6, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-81096-0.2MG
 View more versions of this product
Product Code. 18256269
  • ONLINE EXCLUSIVE PRICE
    € 583.00
    € 551.25 / 0.20mg
    Save € 31.75
    5% Off
Description
CD81 Monoclonal specifically detects CD81 in Human, Goat, Chimpanzee, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Functional.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD81
Monoclonal
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Functional
CD81 antigen, CD81 antigen (target of antiproliferative 1), CD81 molecule, CVID6, TAPA1, TAPA-1, Target of the antiproliferative 1, Tetraspanin-28, Tspan-28, tspan-28,26 kDa cell surface protein TAPA-1, TSPAN28S5.7
Rabbit
Protein A purified
RUO
Primary
This antibody is specific for human CD81, which is expressed on most tissues with the exception of red blood cells and platelets.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG κ
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
1D6
Unconjugated
PBS
CD81
This antibody was produced by immunizing mice with 5A6-aggregated OCI-LY8 B cells.
0.2 mg
Adaptive Immunity, Cell Cycle and Replication, Immunology
975
Human, Chimpanzee, Goat, Sheep