Learn More
CD90 / Thy1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27770-1-AP-150UL
 View more versions of this product
Product Code. 18054475
  • € 410.00 / 150µL
Description
The CD90 / Thy1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD90 / Thy1 . This antibody recognizes human, mouse, rat, pig antigen. The CD90 / Thy1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CD90 / Thy1
450 μg/mL
Rabbit
Primary
Human, Mouse, Rat, Pig
150 kDa
IgG
Western Blot, ELISA
CD90, CD90 / Thy1 , CDw90, Thy 1 antigen, Thy 1 cell surface antigen, Thy 1 membrane glycoprotein, THY1
150 μL
7070
Store at -20°C. Stable for one year after shipment.