missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD90 / Thy1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CD90 / Thy1 |
|---|---|
| Concentration | 450 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066875
|
Proteintech
27770-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18054475
|
Proteintech
27770-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CD90 / Thy1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD90 / Thy1 . This antibody recognizes human, mouse, rat, pig antigen. The CD90 / Thy1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CD90 / Thy1 | |
| Western Blot, ELISA | |
| Rabbit | |
| 7070 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 450 μg/mL | |
| Liquid | |
| Human, Mouse, Rat, Pig | |
| CD90, CD90 / Thy1 , CDw90, Thy 1 antigen, Thy 1 cell surface antigen, Thy 1 membrane glycoprotein, THY1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title