missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc27 Antibody (AF 3.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2606-0.025mg
This item is not returnable.
View return policy
Description
Cdc27 Monoclonal specifically detects Cdc27 in Mammal, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.
Specifications
| Cdc27 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| ANAPC3CDC27 homolog, anaphase promoting complex subunit 3, Anaphase-promoting complex subunit 3, anaphase-promoting complex, protein 3, APC 3, APC3cell division cycle 27, CDC27Hs, cell division cycle 27 homolog (S. cerevisiae), cell division cycle protein 27 homolog, D0S1430E, D17S978E, HNUC, H-NUC, NUC2, nuc2 homolog | |
| Mouse | |
| 80 kDa | |
| 0.025 mg | |
| Cell Cycle and Replication | |
| 996 | |
| Mammalia | |
| Purified |
| Western Blot | |
| AF 3.1 | |
| Unconjugated | |
| P30260 | |
| CDC27 | |
| A peptide corresponding to the C-terminal 10 residues of human cdc27 coupled to KLH. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Cdc27 (AF3.1). Cross-reacts with Human, Mouse, Rat, Xenopus laevis, Cow and Dog. Not yet tested in other species. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction