missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc27 Antibody (AF 3.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00
Specifications
| Antigen | Cdc27 |
|---|---|
| Clone | AF 3.1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot |
Description
Cdc27 Monoclonal specifically detects Cdc27 in Mammal, Xenopus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.Specifications
| Cdc27 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| P30260 | |
| 996 | |
| A peptide corresponding to the C-terminal 10 residues of human cdc27 coupled to KLH. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 80 kDa |
| AF 3.1 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mammalia | |
| ANAPC3CDC27 homolog, anaphase promoting complex subunit 3, Anaphase-promoting complex subunit 3, anaphase-promoting complex, protein 3, APC 3, APC3cell division cycle 27, CDC27Hs, cell division cycle 27 homolog (S. cerevisiae), cell division cycle protein 27 homolog, D0S1430E, D17S978E, HNUC, H-NUC, NUC2, nuc2 homolog | |
| CDC27 | |
| IgG2b | |
| Protein A purified | |
| Cdc27 (AF3.1). Cross-reacts with Human, Mouse, Rat, Xenopus laevis, Cow and Dog. Not yet tested in other species. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title