missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc6, p Ser54 Antibody (SR2085), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21570-100ul
This item is not returnable.
View return policy
Description
Cdc6 Monoclonal antibody specifically detects Cdc6 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| Cdc6 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR2085 | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| CDC18LHsCDC18, Cdc18-related protein, CDC6 (cell division cycle 6, S. cerevisiae) homolog, CDC6 cell division cycle 6 homolog, CDC6 cell division cycle 6 homolog (S. cerevisiae), CDC6-related protein, cell division control protein 6 homolog, cell division cycle 6 homolog (S. cerevisiae), cell division cycle 6 protein, HsCdc18, HsCDC6, p62(cdc6) | |
| A synthesized peptide derived from human phospho-Cdc6 [Ser54] (Uniprot #: Q99741) | |
| 100 μg | |
| Cell Cycle and Replication, Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
| 990 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction