missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc6, p Ser54 Antibody (SR2085), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Cdc6 |
|---|---|
| Clone | SR2085 |
| Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18610224
|
Novus Biologicals
NBP3-21570-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650555
|
Novus Biologicals
NBP3-21570-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cdc6 Monoclonal antibody specifically detects Cdc6 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| Cdc6 | |
| Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CDC18LHsCDC18, Cdc18-related protein, CDC6 (cell division cycle 6, S. cerevisiae) homolog, CDC6 cell division cycle 6 homolog, CDC6 cell division cycle 6 homolog (S. cerevisiae), CDC6-related protein, cell division control protein 6 homolog, cell division cycle 6 homolog (S. cerevisiae), cell division cycle 6 protein, HsCdc18, HsCDC6, p62(cdc6) | |
| A synthesized peptide derived from human phospho-Cdc6 [Ser54] (Uniprot #: Q99741) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR2085 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, Phospho Specific, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 990 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title