Learn More
CDCA5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29316-1-AP-20UL
 View more versions of this product
Product Code. 18044775
  • € 145.00 / 20µL
Description
The CDCA5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CDCA5. This antibody recognizes human antigen. The CDCA5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CDCA5
600 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
CDCA5, p35, SORORIN
20 μL
113130
Store at -20°C. Stable for one year after shipment.