missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCA5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CDCA5 |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18044775
|
Proteintech
29316-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18058085
|
Proteintech
29316-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The CDCA5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CDCA5. This antibody recognizes human antigen. The CDCA5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CDCA5 | |
| Western Blot, ELISA | |
| Rabbit | |
| 113130 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 600 μg/mL | |
| Liquid | |
| Human | |
| CDCA5, p35, SORORIN | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title