Learn More
CDCA5 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29316-1-AP-150UL
This item is not returnable.
View return policy
Description
The CDCA5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CDCA5. This antibody recognizes human antigen. The CDCA5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CDCA5 | |
| 600 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| CDCA5, p35, SORORIN | |
| 150 μL | |
| 113130 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |