Learn More
CDK5 Mouse, anti-Human, Clone: 1B9E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68514-1-IG-150UL
 View more versions of this product
Product Code. 18099675
  • € 410.00 / 150µL
Description
The CDK5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK5. This antibody recognizes human, mouse antigen. The CDK5 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CDK5
1B9E1
CDK5, Cell division protein kinase 5, cyclin dependent kinase 5, PSSALRE, TPKII catalytic subunit
150 μL
1020
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunohistochemistry, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse
150 kDa