Learn More
CDK5 Mouse, anti-Human, Clone: 1B9E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68514-1-IG-150UL
This item is not returnable.
View return policy
Description
The CDK5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK5. This antibody recognizes human, mouse antigen. The CDK5 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| CDK5 | |
| 1B9E1 | |
| CDK5, Cell division protein kinase 5, cyclin dependent kinase 5, PSSALRE, TPKII catalytic subunit | |
| 150 μL | |
| 1020 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG1 |