missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDK5 Mouse, anti-Human, Clone: 1B9E1, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CDK5 |
|---|---|
| Clone | 1B9E1 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18099675
|
Proteintech
68514-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18077905
|
Proteintech
68514-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The CDK5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CDK5. This antibody recognizes human, mouse antigen. The CDK5 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spezifikation
| CDK5 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse | |
| CDK5, Cell division protein kinase 5, cyclin dependent kinase 5, PSSALRE, TPKII catalytic subunit | |
| Primary |
| 1B9E1 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Mouse | |
| 1020 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts