missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11611-2-AP-150UL
This item is not returnable.
View return policy
Description
Using anti-Yo antibodies, three genes have been cloned (CDR 1-3). CDR2 suppresses the transcriptional activity and DNA binding of nuclear factor-kappa B by an unknown mechanism. The neuron specific expression of CDR2 is regulated by a post-transcriptional mechanism.Specifications
| CDR2 | |
| Polyclonal | |
| Unconjugated | |
| CDR2 | |
| CDR2, CDR62, PCD17, Yo | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1039, 12585, 308958 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97817, Q01850 | |
| CDR2 | |
| CDR2 Fusion Protein Ag2183 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction