missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CDR2 |
|---|---|
| Concentration | 0.4 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858803
|
Proteintech
11611-2-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848803
|
Proteintech
11611-2-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Using anti-Yo antibodies, three genes have been cloned (CDR 1-3). CDR2 suppresses the transcriptional activity and DNA binding of nuclear factor-kappa B by an unknown mechanism. The neuron specific expression of CDR2 is regulated by a post-transcriptional mechanism.Specifications
| CDR2 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| P97817, Q01850 | |
| 1039, 12585, 308958 | |
| CDR2 Fusion Protein Ag2183 | |
| Primary | |
| -20°C | |
| CDR2 |
| 0.4 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| CDR2, CDR62, PCD17, Yo | |
| CDR2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title