missing translation for 'onlineSavingsMsg'
Learn More

CEACAM6/CD66c Antibody (1H7-4B) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-54626-0.025mg

 View more versions of this product

Product Code. 18616937

  • € 214.00 / 0.03mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CEACAM6/CD66c Monoclonal antibody specifically detects CEACAM6/CD66c in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CEACAM6/CD66c
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1 - 1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:500, CyTOF-ready
carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
Recombinant human CEACAM6/CD66c extracellular domain.
0.025 mg
Cancer
4680
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
1H7-4B
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.