missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM6/CD66c Antibody (1H7-4B) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 523.00
Specifications
| Antigen | CEACAM6/CD66c |
|---|---|
| Clone | 1H7-4B |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1 - 1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616937
|
Novus Biologicals
NBP2-54626-0.025mg |
0.025 mg |
€ 214.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653969
|
Novus Biologicals
NBP2-54626 |
0.1 mg |
€ 523.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEACAM6/CD66c Monoclonal antibody specifically detects CEACAM6/CD66c in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOFSpecifications
| CEACAM6/CD66c | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS | |
| 4680 | |
| IgG1 κ | |
| Protein A purified |
| 1H7-4B | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1 - 1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen | |
| Recombinant human CEACAM6/CD66c extracellular domain. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title