missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CELA2A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5145306

 View more versions of this product

Product Code. 17970353

  • € 189.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Like most of the human elastases, elastase 2A is secreted from the pancreas as a zymogen. In other species, elastase 2A has been shown to preferentially cleave proteins after leucine, methionine, and phenylalanine residues.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CELA2A
Polyclonal
Unconjugated
CELA2A
CELA2A; chymotrypsin like elastase family member 2A; chymotrypsin-like elastase family member 2A; chymotrypsin-like elastase family, member 2A; Ela2; Ela-2; Ela2a; ELAII; Elaii1; elastase 2; Elastase 2, pancreatic; elastase 2A; elastase II; Elastase-2; elastase-2A; neutrophilic elastase; pancreatic elastase 2; pancreatic elastase IIA; PE-1; preproelastase II
Rabbit
Antigen affinity chromatography, Protein A
RUO
13706, 24332
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
P00774, P05208
CELA2A
Recombinant Mouse ELA2A protein, Cys17-Asn271 (Accession #P05208).
20 μL
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.