missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CELA2A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
€ 185.00 - € 405.00
Specifications
| Antigen | CELA2A |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17970353
|
Invitrogen™
PA5145306 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17900773
![]() |
Invitrogen™
PA5145305 |
200 μL |
€ 405.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Like most of the human elastases, elastase 2A is secreted from the pancreas as a zymogen. In other species, elastase 2A has been shown to preferentially cleave proteins after leucine, methionine, and phenylalanine residues.Specifications
| CELA2A | |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| P00774, P05208 | |
| 13706, 24332 | |
| Recombinant Mouse ELA2A protein, Cys17-Asn271 (Accession #P05208). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| CELA2A; chymotrypsin like elastase family member 2A; chymotrypsin-like elastase family member 2A; chymotrypsin-like elastase family, member 2A; Ela2; Ela-2; Ela2a; ELAII; Elaii1; elastase 2; Elastase 2, pancreatic; elastase 2A; elastase II; Elastase-2; elastase-2A; neutrophilic elastase; pancreatic elastase 2; pancreatic elastase IIA; PE-1; preproelastase II | |
| CELA2A | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| CELA2A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title