missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP135 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05666-20ul
This item is not returnable.
View return policy
Description
CEP135 Polyclonal antibody specifically detects CEP135 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CEP135 | |
| Polyclonal | |
| Western Blot 1:200-1:2000 | |
| centrosomal protein 135kDa, Centrosomal protein 4centrosomal protein of 135 kDa, Cep135, CEP4, FLJ13621, KIAA0635centrosome protein cep135 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 841-1140 of human CEP135 (NP_079285.2). LEAAVQEKEEMKSRVHKYITEVSRWESLMAAKEKENQDLLDRFQMLHNRAEDWEVKAHQAEGESSSVRLELLSIDTERRHLRERVELLEKEIQEHINAHHAYESQISSMAKAMSRLEEELRHQEDEKATVLNDLSSLRELCIKLDSGKDIMTQQLNSKNLEFERVVVELENVKSESDLLKKQLSNERHTVKNLESLLATNRDKEFHSHLTSHEKDTEIQLLKEKLTLSESKLTSQSRENTMLRAKVAQLQTDYDALKRQISTERYERERAIQEMRRHGLATPPLSSTLRSPSHSPEHRNV | |
| 20 μg | |
| Cell Biology, Cell Cycle and Replication | |
| 9662 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction