missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP135 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 462.00
Specifications
| Antigen | CEP135 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CEP135 Polyclonal antibody specifically detects CEP135 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CEP135 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cell Cycle and Replication | |
| PBS with 50% glycerol, pH7.3. | |
| 9662 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| centrosomal protein 135kDa, Centrosomal protein 4centrosomal protein of 135 kDa, Cep135, CEP4, FLJ13621, KIAA0635centrosome protein cep135 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 841-1140 of human CEP135 (NP_079285.2). LEAAVQEKEEMKSRVHKYITEVSRWESLMAAKEKENQDLLDRFQMLHNRAEDWEVKAHQAEGESSSVRLELLSIDTERRHLRERVELLEKEIQEHINAHHAYESQISSMAKAMSRLEEELRHQEDEKATVLNDLSSLRELCIKLDSGKDIMTQQLNSKNLEFERVVVELENVKSESDLLKKQLSNERHTVKNLESLLATNRDKEFHSHLTSHEKDTEIQLLKEKLTLSESKLTSQSRENTMLRAKVAQLQTDYDALKRQISTERYERERAIQEMRRHGLATPPLSSTLRSPSHSPEHRNV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title