Description
- Wide mouth, amber glass
- Cleaned per EPA guidelines for semi-volatiles, pesticides, PCBs and metals
- With white polypropylene cap with PTFE faced foamed polyethylene liner
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Cleaning Procedure | EPA guidelines |
| Color | Amber |
| Material | Glass |
| Quantity | 12/Pk. |
| Closure Color | White |
| Liner Material | PTFE |
| Certifications/Compliance | EPA |
| Product Type | Wide Mouth Bottle |
| Closure Liner | Polyethylene |
| Show More |