missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFAP61 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92398-0.02ml
This item is not returnable.
View return policy
Description
CFAP61 Polyclonal antibody specifically detects CFAP61 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CFAP61 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| chromosome 20 open reading frame 26, dJ1002M8.3, dJ1178H5.4, DKFZp434K156 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CFAP61 (NP_056400.3). MSVLTSPRGKVEVVHCRRTESQDVYCIKSLIRKFTCKLFGKLNIIYLLEKANLAVTLCNDKEEIMAQATFLDYPNWNVAKQDDWVSVFRELDSDIPCTPL | |
| 0.02 mL | |
| Cell Biology | |
| 26074 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction