missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CFAP61 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 231.00 - € 468.00
Specifications
| Antigen | CFAP61 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CFAP61 Polyclonal antibody specifically detects CFAP61 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| CFAP61 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS with 50% glycerol, pH7.3. | |
| 26074 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| chromosome 20 open reading frame 26, dJ1002M8.3, dJ1178H5.4, DKFZp434K156 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human CFAP61 (NP_056400.3). MSVLTSPRGKVEVVHCRRTESQDVYCIKSLIRKFTCKLFGKLNIIYLLEKANLAVTLCNDKEEIMAQATFLDYPNWNVAKQDDWVSVFRELDSDIPCTPL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title