missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92285-0.02ml
This item is not returnable.
View return policy
Description
CHCHD1 Polyclonal antibody specifically detects CHCHD1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CHCHD1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C10orf34, C2360, chromosome 10 open reading frame 34, coiled-coil-helix-coiled-coil-helix domain containing 1, coiled-coil-helix-coiled-coil-helix domain-containing protein 1, FLJ25854, Nuclear protein C2360 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human CHCHD1 (NP_976043.1). MATPSLRGRLARFGNPRKPVLKPNKPLILANRVGERRREKGEATCITEMSVMMACWKQNEFRDDACRKEIQGFLDCAARAQEARKMRSIQETLGESGSLLPNKLNKLLQRFPNKPYLS | |
| 0.02 mL | |
| Cell Biology | |
| 118487 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction