missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 463.00
Specifications
| Antigen | CHCHD1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677651
|
Novus Biologicals
NBP2-92285-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18673981
|
Novus Biologicals
NBP2-92285-0.1ml |
0.1 mL |
€ 463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHCHD1 Polyclonal antibody specifically detects CHCHD1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| CHCHD1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS with 50% glycerol, pH7.3. | |
| 118487 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| C10orf34, C2360, chromosome 10 open reading frame 34, coiled-coil-helix-coiled-coil-helix domain containing 1, coiled-coil-helix-coiled-coil-helix domain-containing protein 1, FLJ25854, Nuclear protein C2360 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human CHCHD1 (NP_976043.1). MATPSLRGRLARFGNPRKPVLKPNKPLILANRVGERRREKGEATCITEMSVMMACWKQNEFRDDACRKEIQGFLDCAARAQEARKMRSIQETLGESGSLLPNKLNKLLQRFPNKPYLS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title