missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD5 Mouse anti-Human, Clone: 2G10A7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66584-1-IG-20UL
This item is not returnable.
View return policy
Description
CHCHD5 (Coiled-Coil-Helix-Coiled-Coil-Helix Domain Containing 5) is a Protein Coding gene. Among its related pathways are Metabolism of proteins and Mitochondrial protein import.Specifications
| CHCHD5 | |
| Monoclonal | |
| 0.93 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BSY4 | |
| CHCHD5 | |
| CHCHD5 Fusion Protein Ag22534 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2G10A7 | |
| Unconjugated | |
| CHCHD5 | |
| C2orf9, CHCHD5 | |
| Mouse | |
| Protein G | |
| RUO | |
| 84269 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction