missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHCHD5 Mouse anti-Human, Clone: 2G10A7, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CHCHD5 |
|---|---|
| Clone | 2G10A7 |
| Concentration | 0.93 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16848595
|
Proteintech
66584-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16838595
|
Proteintech
66584-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CHCHD5 (Coiled-Coil-Helix-Coiled-Coil-Helix Domain Containing 5) is a Protein Coding gene. Among its related pathways are Metabolism of proteins and Mitochondrial protein import.Specifications
| CHCHD5 | |
| 0.93 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C2orf9, CHCHD5 | |
| CHCHD5 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2G10A7 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BSY4 | |
| 84269 | |
| CHCHD5 Fusion Protein Ag22534 | |
| Primary | |
| -20°C | |
| CHCHD5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title