Learn More
Honeywell™ Check and Go DeepTril 5 Gloves
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
Description
- Black HPPE/composite fiber knitted glove
- Fully dipped black foam nitrile coating
- Elasticated knitted wrist
- The ergonomics seamless knit construction conforms better to the natural shape of the hand for greater strength and durability, superior confort
- The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The foam coating optimise the glove's breathability
- The nitrile foam and PVC dots on palm improves the grip
- The knitted and elasticated wrist gives the glove good support
- The nitrile foam coating provides outstanding abrasion resistance and good protection in light oily environnement
- HPPE mixed with composite fiber provides very high resistance to cut
- The black color is suitable for work in a soiled atmosphere
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 Basic standards, EN 388-2016 Mechanical hazards |
| Description | 13 gauge |
| Color | Black |
| Material | Nitrile, HPPE |
| Size | 8 |
| Length (Metric) | 25 cm |