Learn More
Honeywell™ Check and Go DeepTril 5 Gloves
Product Code. 15876755
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
Product Code. 15876755
Brand: Honeywell™ 229940509
Only null left
Add to basket
Add to basket
Honeywell™ Check and Go DeepTril 5 Gloves are fully dipped in a nitrile foam coating for handling in oily or greasy conditions.
  • Black HPPE/composite fiber knitted glove
  • Fully dipped black foam nitrile coating
  • Elasticated knitted wrist
  • The ergonomics seamless knit construction conforms better to the natural shape of the hand for greater strength and durability, superior confort
  • The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
  • The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The foam coating optimise the glove's breathability
  • The nitrile foam and PVC dots on palm improves the grip
  • The knitted and elasticated wrist gives the glove good support
  • The nitrile foam coating provides outstanding abrasion resistance and good protection in light oily environnement
  • HPPE mixed with composite fiber provides very high resistance to cut
  • The black color is suitable for work in a soiled atmosphere
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-2003 Basic standards, EN 388-2016 Mechanical hazards
Description 13 gauge
Color Black
Material Nitrile, HPPE
Size 9
Length (Metric) 25 cm