Learn More
Schneider Gerd™ Chemical Shovel
Use these scoops for a variety of applications. Schneider Gerd™ Chemical Shovels are available in cast aluminum and stainless steel.
Brand:  Schneider Gerd™ 526
Additional Details : Weight : 0.15300kg
 View more versions of this product
Product Code. 10602635
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Metal lab scoops
  • For transferring or weighing solids
Specifications
Specifications
Chemical Shovel
290 x 105 mm
105 mm
Cast Aluminum
290 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers