missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CHIC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
€ 618.00
Specifications
| Antigen | CHIC2 |
|---|---|
| Concentration | 0.1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17108853
|
Invitrogen™
PA5110926 |
100 μL |
€ 618.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17189653
![]() |
Invitrogen™
PA5111741 |
100 μL |
€ 618.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: SIEKLLEWEN NRLYHKLCLH WRLSKRKCET N Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
This gene encodes a member of the CHIC family of proteins. The encoded protein contains a cysteine-rich hydrophobic (CHIC) motif, and is localized to vesicular structures and the plasma membrane. This gene is associated with some cases of acute myeloid leukemia.Specifications
| CHIC2 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| CHIC2 | |
| 1700081B18Rik; 4930502K01Rik; BrX-like translocated in leukemia; BTL; Chic2; cysteine rich hydrophobic domain 2; cysteine-rich hydrophobic domain 2; cysteine-rich hydrophobic domain 2 protein; cysteine-rich hydrophobic domain-containing protein 2; cystein-rich hydrophobic domain 2; MGC21173; zgc:55670 | |
| CHIC2 | |
| Primary | |
| Antigen affinity chromatography |
| 0.1 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKJ5 | |
| 26511 | |
| Recombinant Protein corresonding to Human CHIC2. Recombinant protein control fragment (Product #RP-90088). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title