missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethyl-n-octylsilane, 97%, Thermo Scientific Chemicals

Catalog Number 11359235
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
18162-84-0
C10H23ClSi
206.829
MFCD00000511
DBKNGKYVNBJWHL-UHFFFAOYSA-N
n-octyldimethylchlorosilane, chlorodimethyloctylsilane, octyldimethylchlorosilane, chlorodimethyl octyl silane, silane, chlorodimethyloctyl, chloro dimethyl octylsilane, dimethyloctylchlorosilane, dimethyl-n-octylchlorosilane, dimethyloctylsilyl chloride, acmc-1bvuc
87482
chloro-dimethyl-octylsilane
CCCCCCCC[Si](C)(C)Cl
This item is not returnable. View return policy
18162-84-0
C10H23ClSi
206.829
MFCD00000511
DBKNGKYVNBJWHL-UHFFFAOYSA-N
n-octyldimethylchlorosilane, chlorodimethyloctylsilane, octyldimethylchlorosilane, chlorodimethyl octyl silane, silane, chlorodimethyloctyl, chloro dimethyl octylsilane, dimethyloctylchlorosilane, dimethyl-n-octylchlorosilane, dimethyloctylsilyl chloride, acmc-1bvuc
87482
chloro-dimethyl-octylsilane
CCCCCCCC[Si](C)(C)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 222°C to 225°C
Quantity 5 g
UN Number UN2987
Beilstein 1738376
Flash Point 92°C (198°F)
Formula Weight 206.83
Percent Purity 97%
Odor Acrid
Refractive Index 1.435
Sensitivity Moisture sensitive
Density 0.873
Chemical Name or Material Chlorodimethyl-n-octylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'