Learn More
Cholic acid, MedChemExpress
Product Code. 16419056
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419056 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16419056 Supplier Medchem Express Supplier No. HYN0324
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MedChemExpress Cholic acid is a major primary bile acid produced in the liver and usually conjugated with glycine or taurine. It facilitates fat absorption and cholesterol excretion. Cholic acid is orally active.
Cholic acid is a major primary bile acid produced in the liver and usually conjugated with glycine or taurine. It facilitates fat absorption and cholesterol excretion. Cholic acid is orally active.
Specifications
Chemical Name or Material Cholic acid
CAS 81-25-4
Color White
Purity Grade Notes Research
Molecular Formula C24H40O5
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (122.38 mM) ∣H2O : 0.67 mg/mL (1.64 mM; Need ultrasonic)
SMILES C[C@H](CCC(O)=O)[C@@]1([H])CC[C@@]2([H])[C@]3([H])[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@@]21C
Molecular Weight (g/mol) 408.57
Formula Weight 408.57
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less
Research purposes only