missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin A Mouse, Clone: LK2H10 + PHE5 + CGA/414, Novus Biologicals™
Mouse Monoclonal Antibody
€ 295.00 - € 514.00
Specifications
| Form | Purified |
|---|---|
| Target Species | Pig |
| Isotype | IgG |
| Content And Storage | Store at 4C. |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18797023
|
Novus Biologicals
NBP2-34240-0.02mg |
€ 295.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18154102
|
Novus Biologicals
NBP2-34240-0.1MG |
0.1 mg |
€ 514.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromogranin A Monoclonal specifically detects Chromogranin A in Human, Mouse, Rat, Porcine, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Purified | |
| Pig | |
| IgG | |
| Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title