Learn More
CINP Monoclonal Antibody (OTI8E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812290
This item is not returnable.
View return policy
Description
The function of this gene has not been reported. This gene contains a trinucleotide (ctt) repeat length polymorphism; alleles with either one or two repeat units have been identified.Specifications
| CINP | |
| Monoclonal | |
| Unconjugated | |
| CINP | |
| cyclin dependent kinase 2 interacting protein | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51550 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI8E4 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BW66 | |
| CINP | |
| Full length human recombit protein of human CINP produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |