missing translation for 'onlineSavingsMsg'
Learn More

CINP Monoclonal Antibody (OTI8E4), TrueMAB™, OriGene

Mouse Monoclonal Antibody

€ 242.00 - € 540.87

Specifications

Antigen CINP
Clone OTI8E4
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17576704
Encompass_Preferred
View Documents
Origene Technologies
TA812290S
30 μL
€ 242.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17556704
Encompass_Preferred
View Documents
Origene Technologies
TA812290
100 μL
€ 540.87
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CINP Monoclonal antibody specifically detects CINP in Human samples. It is validated for Western Blot
Specifications

Specifications

CINP
Western Blot
Unconjugated
Mouse
Human
51550
Primary
CINP
OTI8E4
Monoclonal
Liquid
RUO
Q9BW66
IgG2b
-20° C, Avoid Freeze/Thaw Cycles
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.