Learn More
CINP Monoclonal Antibody (OTI8E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812290S
This item is not returnable.
View return policy
Description
CINP Monoclonal antibody specifically detects CINP in Human samples. It is validated for Western BlotSpecifications
| CINP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BW66 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51550 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI8E4 | |
| Unconjugated | |
| CINP | |
| 1810047K05Rik; 2810452K22Rik; CDK2-interacting protein; Cinp; cyclin dependent kinase 2 interacting protein; cyclin-dependent kinase 2 interacting protein; cyclin-dependent kinase 2-interacting protein; MGC849; RGD1563395 | |
| Full length human recombinant protein of human CINP produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |