Learn More
CINP Monoclonal Antibody (OTI8E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA812290S
Encompass_Preferred
 View more versions of this product
Product Code. 17576704
  • € 242.00 / 30µL
Description
CINP Monoclonal antibody specifically detects CINP in Human samples. It is validated for Western Blot
Specifications
Specifications
CINP
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BW66
Mouse
Affinity chromatography
RUO
51550
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI8E4
Unconjugated
CINP
1810047K05Rik; 2810452K22Rik; CDK2-interacting protein; Cinp; cyclin dependent kinase 2 interacting protein; cyclin-dependent kinase 2 interacting protein; cyclin-dependent kinase 2-interacting protein; MGC849; RGD1563395
Full length human recombinant protein of human CINP produced in HEK293T cell.
30 μL