Learn More
cis-Stilbene, 97%
Product Code. p-7081776
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11364549
Brand: Thermo Scientific Alfa Aesar A11924.03
Request Bulk or Custom Format
CAS: 645-49-8 | C14H12 | 180.25 g/mol
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-Stilbene is used as a phosphor and a scintillator. It is mainly used in the produciton of dyes and optical brighteners.
Solubility
Immiscible with water. Miscible with ethanol.
Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 645-49-8
Molecular Formula C14H12
Molecular Weight (g/mol) 180.25
MDL Number MFCD00004788
InChI Key PJANXHGTPQOBST-QXMHVHEDSA-N
Synonym cis-stilbene, z-stilbene, z-1,2-diphenylethene, cis-1,2-diphenylethylene, isostilbene, cis-diphenylethene, stilbene, z, z-2-phenylethenyl benzene, benzene, 1,1'-1,2-ethenediyl bis-, z, z-1,2-diphenylethylene
PubChem CID 5356785
ChEBI CHEBI:36008
IUPAC Name (Z)-stilbene
SMILES C1=CC=C(C=C1)C=CC2=CC=CC=C2
Specifications
Melting Point 1°C to 2°C
Density 1.04
Boiling Point 307°C
Flash Point >110°C (230°F)
Odor Weak
Refractive Index 1.622
Quantity 1 g
Beilstein 1616739
Sensitivity Light sensitive
Merck Index 14,8817
Solubility Information Immiscible with water. Miscible with ethanol.
Formula Weight 180.25
Percent Purity 97%
Chemical Name or Material cis-Stilbene