Learn More
Cisplatin, TRC
Product Code. 18092730
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
missing translation for 'unitSize'
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity unitSize
18092730 100 mg 100mg
18012840 50 mg 50mg
18032740 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 18092730 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC499500100MG
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 15663-27-1
Molecular Formula Cl2 H6 N2 Pt
Molecular Weight (g/mol) 300.04
Synonym Cisplatin, cis-Diamminedichloroplatinum(II)
IUPAC Name azanide;dichloroplatinum(2+)
SMILES [Cl-][Pt+2]([Cl-])([NH3])[NH3]
Spécification
Quantity 100 mg
InChI Formula InChI=1S/2ClH.2H3N.Pt/h2*1H;2*1H3;/q;;;;+2/p-2
Formula Weight 298.9556
Chemical Name or Material Cisplatin