missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-16 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92588-0.02ml
This item is not returnable.
View return policy
Description
Claudin-16 Polyclonal antibody specifically detects Claudin-16 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Claudin-16 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, Flow Cytometry | |
| claudin 16, claudin-16, HOMG3paracellin-1, Paracellin-1, PCLN-1, PCLN1hypomagnesemia 3, with hypercalciuria and nephrocalcinosis | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human CLDN16 (NP_006571.1). HLSGARAGVCPCCHPDGLLATMRDLLQYIACFFAFFSAGFLIVATWTDCWMVNADDSLEVSTKCRGLWWECVTNAFDGIRTCDEYDSILAEHPLKLVVTRA | |
| 0.02 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 10686 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction