missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-16 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | Claudin-16 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Flow Cytometry |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Claudin-16 Polyclonal antibody specifically detects Claudin-16 in Human, Mouse samples. It is validated for Western Blot, Flow CytometrySpecifications
| Claudin-16 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 10686 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, Flow Cytometry | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| claudin 16, claudin-16, HOMG3paracellin-1, Paracellin-1, PCLN-1, PCLN1hypomagnesemia 3, with hypercalciuria and nephrocalcinosis | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human CLDN16 (NP_006571.1). HLSGARAGVCPCCHPDGLLATMRDLLQYIACFFAFFSAGFLIVATWTDCWMVNADDSLEVSTKCRGLWWECVTNAFDGIRTCDEYDSILAEHPLKLVVTRA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title