Learn More
Phenomenex™ Cleanert™ Crimp Caps with Septa, 11 mm, No Slit
Product Code. 31744781
Change view
Click to view available options
Septum Color:
Red/White
White/Red
Unit Size:
Each
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Septum Color unitSize
31744781 White/Red Pack of 100
31745776 Red/White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31744781 Supplier Phenomenex™ Supplier No. AV61100
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
Accessories for use with chromatography vials and in various chromatography applications.
  • Brand: Cleanert
  • Technique: Accessory
TRUSTED_SUSTAINABILITY
Specifications
Color Silver
Diameter (Metric) Outer 11 mm
Material Aluminum
Pre-slit No
Sterility Sterile
Septum PTFE/Silicone
Closure Size 11 mm
Marking Spot No
Autoclavable Autoclavable
Product Type Cap
Septum Color White/Red