Learn More
Fisherbrand™ Clear Borosilicate Glass Volumetric Flasks
Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
10 mL
50 mL
500 mL
Unit Size:
Pack of 2
Pack of 5
Description
- Graduated
- Polyethylene snap cap
- Clear
Specifications
Specifications
| Closure Material | Polyethylene |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass |
| No. per Case | 2 |
| Graduated | Yes |
| Mouth Style | Beaded |
| Class | A |
| Closure Type | Snap |