Learn More
Fisherbrand™ Clear Straight Sided Polystyrene Jars with White Polypropylene Caps
Product Code. p-7144780 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
30 mL
60 mL
120 mL
240 mL
480 mL
950 mL
Unit Size:
Pack of 24
Pack of 36
Pack of 48
Pack of 72
Product Code. 16144457
Only null left
Add to basket
Add to basket
Wide mouth design is great for large solid samples and allow for complete removal of contents
  • Features wide neck and straight sides to facilitate easy recovery of contents
  • Rigid clear polystyrene jars offer glass-like clarity with a slip-resistant smooth surface
Specifications
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Clear
Diameter (Metric) Outer Neck 39 mm
Diameter (Metric) Inner Neck 37 mm
For Use With (Application) Large Solid Samples and Allow for Complete Removal of Contents
Includes Cap
Height (Metric) With Closure 44 mm
Material Polystyrene
Quantity 72
Closure Color White
Neck Finish 43-400
Clarity Clear
Closure Included Yes
Mouth Wide
Shape Round
Height (Metric) 41 mm
Product Type Straight-sided Jar
Closure Liner Polyethylene Foam
Closure Material Polypropylene
Closure Size 43-400 mm
Diameter (Metric) 39 mm
Closure Type Cap