missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIP4 Antibody (OTI5D10), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
€ 256.00 - € 550.00
Specifications
| Antigen | CLIP4 |
|---|---|
| Clone | OTI5D10 |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:500 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306285
|
Novus Biologicals
NBP3-44957-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306731
|
Novus Biologicals
NBP3-44957-30ul |
30 μL |
€ 256.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CLIP4 Monoclonal antibody specifically detects CLIP4 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| CLIP4 | |
| Western Blot 1:2000, Immunohistochemistry 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 79745 | |
| IgG1 | |
| >95% | |
| Antibody |
| OTI5D10 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| CAP-GLY domain containing linker protein family, member 4, CAP-Gly domain-containing linker protein 4, FLJ21069, FLJ32705, restin-like 2, Restin-like protein 2, RSNL2 | |
| Full length human recombinant protein of human CLIP4(NP_078968) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title