missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLN3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92767-0.02ml
This item is not returnable.
View return policy
Description
CLN3 Polyclonal antibody specifically detects CLN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CLN3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Batten disease protein, Batten, Spielmeyer-Vogt disease, battenin, BTSMGC102840, ceroid-lipofuscinosis, neuronal 3, JNCL, Protein CLN3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CLN3 (NP_001035897.1). MGGCAGSRRRFSDSEGEETVPEPRLPLLDHQGAHWKNAVGFWLLGLCNNFSYVVMLSAAHDILSHKRTSGNQSHVDPGPTPIPHNSSSRFDCNSVSTAAVLLADILPTLVIKLLAPLGLHLLPYSPRVLVSGICAAGSFVLVAFSHSVGTSLCGVVFASISSGLGEVTFLSLTAFYPRAVISWWSSGTGGAGLLGALSYLGLTQAGLSPQQTLLSMLGIPALLLASYFLLLTSPEAQDPGGEEEAESAARQPLIRTEAPESKPGSSSSLSLRERWTVFKG | |
| 0.02 mL | |
| Apoptosis, Cell Biology, Neurodegeneration, Neuroscience | |
| 1201 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction