missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLN3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | CLN3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CLN3 Polyclonal antibody specifically detects CLN3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| CLN3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology, Neurodegeneration, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 1201 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Batten disease protein, Batten, Spielmeyer-Vogt disease, battenin, BTSMGC102840, ceroid-lipofuscinosis, neuronal 3, JNCL, Protein CLN3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human CLN3 (NP_001035897.1). MGGCAGSRRRFSDSEGEETVPEPRLPLLDHQGAHWKNAVGFWLLGLCNNFSYVVMLSAAHDILSHKRTSGNQSHVDPGPTPIPHNSSSRFDCNSVSTAAVLLADILPTLVIKLLAPLGLHLLPYSPRVLVSGICAAGSFVLVAFSHSVGTSLCGVVFASISSGLGEVTFLSLTAFYPRAVISWWSSGTGGAGLLGALSYLGLTQAGLSPQQTLLSMLGIPALLLASYFLLLTSPEAQDPGGEEEAESAARQPLIRTEAPESKPGSSSSLSLRERWTVFKG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title