missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92059-0.1ml
This item is not returnable.
View return policy
Description
CNOT10 Polyclonal antibody specifically detects CNOT10 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CNOT10 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CCR4-NOT transcription complex subunit 10, CCR4-NOT transcription complex, subunit 10, DKFZp434K115, FLJ12890, FLJ13165 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 465-744 of human CNOT10 (NP_056257.1). LRNALLLLPEEQQDPKQENGAKNSNQLGGNTESSESSETCSSKSHDGDKFIPAPPSSPLRKQELENLKCSILACSAYVALALGDNLMALNHADKLLQQPKLSGSLKFLGHLYAAEALISLDRISDAITHLNPENVTDVSLGISSNEQDQGSDKGENEAMESSGKRAPQCYPSSVNSARTVMLFNLGSAYCLRSEYDKARKCLHQAASMIHPKEVPPEAILLAVYLELQNGNTQLALQIIKRNQLLPAVKTHSEVRKKPVFQPVHPIQPIQMPAFTTVQRK | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 25904 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction